Learn More
Sino Biological Rat C1qR1/CD93 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786307
Encompass_Preferred
The Rat CD93/C1qR ELISA Pair Set is for the quantitative determination of Rat CD93/C1qR.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat CD93/C1qR coated on a 96-well plate. Standards and samples are added to the wells, and any Rat CD93/C1qR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat CD93/C1qR polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat CD93/C1qR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86307 Supplier Sino Biological Supplier No. SEK802075
The Rat CD93/C1qR ELISA Pair Set is for the quantitative determination of Rat CD93/C1qR.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat CD93/C1qR coated on a 96-well plate. Standards and samples are added to the wells, and any Rat CD93/C1qR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat CD93/C1qR polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat CD93/C1qR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.