Learn More
BIOHIPPO Rat C3a Microsample
SDP
Catalog No. 5000315663
Encompass
Rat C3a Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C3a in serum plasma and other biological fluids It provides a detection range of 7 81-500 ng/mL with a sensitivity of 2 01 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat C3a Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat C3a Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat C3a biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15663 Supplier BIOHIPPO Supplier No. BHE15212653
Rat C3a Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C3a in serum plasma and other biological fluids It provides a detection range of 7 81-500 ng/mL with a sensitivity of 2 01 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat C3a Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat C3a Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat C3a biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color