Learn More
Biomatik Corporation Rat C3 (Complement Component 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58703
Encompass_Preferred
Rat C3 (Complement Component 3) ELISA Kit for detecting C3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01026. Target Synonyms: C3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5584