Learn More
Biomatik Corporation Rat C4 (Complement Component 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58562
Encompass_Preferred
Rat C4 (Complement Component 4) ELISA Kit for detecting C4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08649. Target Synonyms: C4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4465