Learn More
ACROBiosystems Rat Cadherin-17 / CDH17 Protein, His Tag (MALS verified)
Catalog No. 502331945
Encompass_Preferred
Rat Cadherin-17 Protein, His Tag (CA7-R52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Met 786 (Accession # P55281-1).
Catalog No. 50-233-1945 Supplier ACROBiosystems Supplier No. CA7R52H3100UG
Rat Cadherin-17 Protein, His Tag (CA7-R52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Met 786 (Accession # P55281-1).