Learn More
Innovive Inc Rat cage bottom for use with Innovive Rat IVC or static Rat lids, prefilled with Alpha-dri bedding.
Catalog No. NC2193839
Encompass
Alpha-Dri bedding is ideal for GLP research. Standard disposable mouse cage bottom arrives pre-bedded and ready for use with ALPHA-dri. Perfect for GLP studies. PRE-BEDDED, ALPHA-DRI
This item is not returnable. View return policy
Catalog No. NC2193839 Supplier Innovive Inc Supplier No. RBTMHAD
Alpha-Dri bedding is ideal for GLP research. Standard disposable mouse cage bottom arrives pre-bedded and ready for use with ALPHA-dri. Perfect for GLP studies. PRE-BEDDED, ALPHA-DRI