Learn More
BIOHIPPO Rat CALB Microsample
SDP
Catalog No. 5000315562
Encompass
Rat CALB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CALB in Serum plasma tissue homogenates urine and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 13 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CALB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CALB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CALB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15562 Supplier BIOHIPPO Supplier No. BHE15212505
Rat CALB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CALB in Serum plasma tissue homogenates urine and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 13 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CALB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CALB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CALB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color