Learn More
Biomatik Corporation Rat Calbindin (CALB) ELISA Kit
Supplier:  Biomatik Corporation EKU02849
Encompass_Preferred
Rat Calbindin (CALB) ELISA Kit for detecting CALB in serum, plasma, tissue homogenates, urine, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 9.38-600pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07171. Gene ID: 83839. Target Synonyms: CAB27; Calbindin 1,28kDa; Calbindin D28; Vitamin D-dependent calcium-binding protein, avian-type. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5841