Learn More
Biomatik Corporation Rat Calreticulin (CALR) CLIA Kit
Supplier:  Biomatik Corporation EKU09791
Encompass_Preferred
Rat Calreticulin (CALR) CLIA Kit for detecting CALR in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.21pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P18418. Gene ID: 64202. Target Synonyms: CRT; RO; SSA; CC1qR; ERp60; HACBP; grp60; CRTC; CRP55; Calregulin; Sicca Syndrome Antigen A; Autoantigen Ro; Endoplasmic reticulum resident protein 60. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4581