Learn More
Biomatik Corporation Rat Calsyntenin 3 (CLSTN3) ELISA Kit
Supplier:  Biomatik Corporation EKU12053
Encompass_Preferred
Rat Calsyntenin 3 (CLSTN3) ELISA Kit for detecting CLSTN3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R553. Gene ID: 171393. Target Synonyms: CSTN3; CDHR14; CS3; Alc-beta; Cadherin-Related Family Member 14; Alcadein-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6923