Learn More
Biomatik Corporation Rat Calumenin (CALU) ELISA Kit
Supplier:  Biomatik Corporation EKU02899
Encompass_Preferred
Rat Calumenin (CALU) ELISA Kit for detecting CALU in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35783. Target Synonyms: Crocalbin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4615