Learn More
Biomatik Corporation Rat Carboxylesterase 1 (CES1) ELISA Kit
Supplier:  Biomatik Corporation EKU02929
Encompass_Preferred
Rat Carboxylesterase 1 (CES1) ELISA Kit for detecting CES1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 14.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63108. Target Synonyms: ACAT; CEH; CES2; HMSE1; PCE1; SES1; TGH; Retinyl ester hydrolase; Serine esterase 1; Triacylglycerol hydrolase; Egasyn; Cocaine carboxylesterase; Liver Carboxylesterase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4639