Learn More
Biomatik Corporation Rat Carboxypeptidase A1, Pancreatic (CPA1) ELISA Kit
Supplier:  Biomatik Corporation EKU02931
Encompass_Preferred
Rat Carboxypeptidase A1, Pancreatic (CPA1) ELISA Kit for detecting CPA1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.53ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00731. Gene ID: 24269. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5926