Learn More
American Research Products Inc Rat Cardiac Troponin I
Catalog No. 5000330231
Encompass
Rat Cardiac Troponin (cTn- ) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30231 Supplier American Research Products Inc Supplier No. CSBE08594R
Rat Cardiac Troponin (cTn- ) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich