Learn More
American Research Products Inc Rat Cardiac Troponin I
Supplier:  American Research Products Inc CSBE08594RIS
Encompass
Rat Cardiac Troponin (cTn- ) ELISA kit from CUSABIO Detection Range 31 25pg/mL-2000pg/mL Sensitivity 15 6pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30232