Learn More
American Research Products Inc Rat Cardiac Troponin T cTn T
Catalog No. 5000329568
Encompass
Rat Cardiac Troponin T (cTn-T) ELISA kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29568 Supplier American Research Products Inc Supplier No. CSBE16443R
Rat Cardiac Troponin T (cTn-T) ELISA kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma tissue homogenates Method Sandwich