Learn More
Biomatik Corporation Rat Cardiotrophin Like Cytokine Factor 1 (CLCF1) ELISA Kit
Supplier:  Biomatik Corporation EKU02957
Encompass_Preferred
Rat Cardiotrophin Like Cytokine Factor 1 (CLCF1) ELISA Kit for detecting CLCF1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4KMB8. Gene ID: 365395. Target Synonyms: CLC; NR6; BSF3; CISS2; NNT1; Novel Neurotrophin-1; B Cell-Stimulating Factor-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7264