Learn More
Biomatik Corporation Rat Carnitine Acetyltransferase (CRAT) ELISA Kit
Supplier:  Biomatik Corporation EKU02959
Encompass_Preferred
Rat Carnitine Acetyltransferase (CRAT) ELISA Kit for detecting CRAT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q704S8. Gene ID: 311849. Target Synonyms: CAT1; Carnitine O-Acetyltransferase; Carnitine acetylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4397