Learn More
American Research Products Inc Rat cartilage oligomeric matri
Catalog No. 5000329363
Encompass
Rat cartilage oligomeric matriX protein COMP ELISA Kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29363 Supplier American Research Products Inc Supplier No. CSBE13833R
Rat cartilage oligomeric matriX protein COMP ELISA Kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich