Learn More
Biomatik Corporation Rat Cartilage Oligomeric Matrix Protein (COMP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11633
Encompass_Preferred
Rat Cartilage Oligomeric Matrix Protein (COMP) Mini sample ELISA Kit for detecting COMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35444. Gene ID: 25304. Target Synonyms: MED; EDM1; EPD1; PSACH; THBS5; TSP-5; Pseudoachondroplasia,Epiphyseal Dysplasia 1,Multiple; Thrombospondin 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8115