Learn More
American Research Products Inc Rat Caspase 1 CASP1 ELISA ki
Catalog No. 5000327132
Encompass
Rat Caspase-1 (CASP1) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell lysates cell culture supernates Method Sandwich
Catalog No. 50-003-27132 Supplier American Research Products Inc Supplier No. CSBEL004543RA
Rat Caspase-1 (CASP1) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell lysates cell culture supernates Method Sandwich