Learn More
Biomatik Corporation Rat Caspase 14 (CASP14) ELISA Kit
Supplier:  Biomatik Corporation EKU02987
Encompass_Preferred
Rat Caspase 14 (CASP14) ELISA Kit for detecting CASP14 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZZ65. Gene ID: 299587. Target Synonyms: Caspase 14 Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6981