Learn More
Cusabio Technology LLC Rat Caspase 3Casp-3 ELISA Kit
Catalog No. 5000050201
Encompass_Preferred
This kit detects CASP3 (Caspase-3) in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 312 ng/mL to 20 ng/mL with a sensitivity of 0 078 ng/mL enabling precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Results are measured at an absorbance wavelength of 450 nm providing reliable data for apoptosis-related research Designed for flexibility it accommodates multiple biological matrices while maintaining robust performance making it suitable for both routine laboratory analysis and experimental studies requiring accurate quantification of Caspase-3 activity
Catalog No. 50-000-50201 Supplier Cusabio Technology LLC Supplier No. CSBE08857R
This kit detects CASP3 (Caspase-3) in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 312 ng/mL to 20 ng/mL with a sensitivity of 0 078 ng/mL enabling precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Results are measured at an absorbance wavelength of 450 nm providing reliable data for apoptosis-related research Designed for flexibility it accommodates multiple biological matrices while maintaining robust performance making it suitable for both routine laboratory analysis and experimental studies requiring accurate quantification of Caspase-3 activity