Learn More
Biomatik Corporation Rat Caspase 7 (CASP7) ELISA Kit
Supplier:  Biomatik Corporation EKU02997
Encompass_Preferred
Rat Caspase 7 (CASP7) ELISA Kit for detecting CASP7 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88550. Gene ID: 64026. Target Synonyms: CMH1; ICE-LAP3; MCH3; Caspase 7 Apoptosis-Related Cysteine Peptidase; Cysteine Aspartate Protease 7; Cysteinyl Aspartate Specific Proteinases 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6571