Learn More
Biomatik Corporation Rat Caspase 8 (CASP8) ELISA Kit
Supplier:  Biomatik Corporation EKU03000
Encompass_Preferred
Rat Caspase 8 (CASP8) ELISA Kit for detecting CASP8 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JHX4. Gene ID: 64044. Target Synonyms: CAP4; FLICE; MACH; MCH5; Cysteinyl Aspartate Specific Proteinases 8; Apoptotic cysteine protease; FADD-homologous ICE/ced-3-like protease; MORT1-associated ced-3 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6572