Learn More
Biomatik Corporation Rat Cathepsin D (CTSD) ELISA Kit
Supplier:  Biomatik Corporation EKU03027
Encompass_Preferred
Rat Cathepsin D (CTSD) ELISA Kit for detecting CTSD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 24.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24268. Target Synonyms: CPSD; CLN10; Lysosomal Aspartyl Protease; Ceroid-Lipofuscinosis,Neuronal 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4574