Learn More
Biomatik Corporation Rat Cathepsin G (CTSG) ELISA Kit
Supplier:  Biomatik Corporation EKU03032
Encompass_Preferred
Rat Cathepsin G (CTSG) ELISA Kit for detecting CTSG in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G3V9Q7. Gene ID: 290257. Target Synonyms: CG; CTS-G. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9070