Learn More
Kingfisher Biotech Inc Rat CCL2 Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1746R100
Encompass
The Rat CCL2 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rat CCL2 applications are for cell culture, ELISA standard, and Western Blot Control. Rat CCL2 yeast-derived recombinant protein can be purchased in multiple sizes. Rat CCL2 Specifications: (Molecular Weight: 14.1 kDa) (Amino Acid Sequence: QPDAVNAPLT CCYSFTGKMI PMSRLENYKR ITSSRCPKEA VVFVTKLKRE ICADPNKEWV QKYIRKLDQN QVRSETTVFY KIASTLRTSA PLNVNLTHKS EANASTLFST TTSSTSVEVT SMTEN (125)) (Gene ID: 24770). For research use only. Made in the USA
Catalog No. 50-253-2837