Learn More
BIOHIPPO Rat CD14 Microsample
SDP
Catalog No. 5000314623
Encompass
Rat CD14 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD14 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 29 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD14 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD14 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD14 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14623 Supplier BIOHIPPO Supplier No. BHE15212116
Rat CD14 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD14 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 29 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD14 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD14 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD14 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color