Learn More
Sino Biological Anti-CD14 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80308RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD14 (rR CD14; Catalog#80308-R08H; Q63691; Met1-Tyr341). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1902