Learn More
Sino Biological Rat CD14 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80308R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD14 (Q63691) (Met1-Tyr341) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3517