Learn More
Sino Biological Rat CD14 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80308R08H100
Encompass_Preferred
A DNA sequence encoding the rat Cd14 (Q63691) (Met1-Tyr341) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2523