Learn More
Sino Biological Anti-CD164 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80354T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD164 / Endolyn (Q9QX82; Met1-Asp160). CD164 / Endolyn specific IgG was purified by Rat CD164 / Endolyn affinity chromatography.
Catalog No. 50-240-8047