Learn More
BIOHIPPO Rat CD1A Microsample
SDP
Catalog No. 5000314967
Encompass
Rat CD1A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD1A in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 07 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD1A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD1A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD1A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14967 Supplier BIOHIPPO Supplier No. BHE15212373
Rat CD1A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD1A in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 07 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD1A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD1A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD1A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color