Learn More
BIOHIPPO Rat CD200 Microsample
SDP
Catalog No. 5000313854
Encompass
Rat CD200 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD200 in Serum plasma and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 18 71 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD200 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD200 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD200 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13854 Supplier BIOHIPPO Supplier No. BHE15210643
Rat CD200 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD200 in Serum plasma and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 18 71 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD200 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD200 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD200 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color