Learn More
ACROBiosystems Rat CD22 Protein, His Tag
Supplier:  ACROBiosystems CD2R52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Rat CD22, His Tag (CD2-R52H3) is expressed from human 293 cells (HEK293). It contains AA Trp 24 - Arg 692 (Accession # D3ZD88-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 77.1 kDa.
Catalog No. 50-205-4726