Learn More
Sino Biological Rat CD24 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80297R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD24 (Q07490) (Met1-Cys55) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2964