Learn More
Sino Biological Rat CD274 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80450R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD274 (NP_001178883.1)(Met1-Thr238) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-0748