Learn More
Sino Biological Rat CD276 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80380R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD276 (Q7TPB4) (Met1-Phe244) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2677