Learn More
ACROBiosystems ACROBiosystems Rat CD3 epsilon&CD3 delta Heterodimer Protein, His Tag&Flag Tag (MALS verified)
Catalog No. 502107349
Encompass_Preferred
Recombinant Protein;100UG;Rat CD3E&CD3D Heterodimer Protein, His Tag&Flag Tag (CDD-R52D8) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Asp 103 (CD3E) & Phe 22 - Gly 106 (CD3D) (Accession # D4A5M2-1 (CD3E) & P19377-1 (CD3D)).;Rat CD3E&CD3D Heterodimer Protein, His Tag&Flag Tag is produced by co-expression of CD3E and CD3D, has a calculated MW of 14.9 kDa (CD3E) and 14.5 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3D is fused with a flag tag at the C-terminus. As a result of glycosylation, the heterodimer protein migrates as 19-20 kDa under reducing (R) condition, and 40-45 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-210-7349 Supplier ACROBiosystems Supplier No. CDDR52D8100UG
Recombinant Protein;100UG;Rat CD3E&CD3D Heterodimer Protein, His Tag&Flag Tag (CDD-R52D8) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Asp 103 (CD3E) & Phe 22 - Gly 106 (CD3D) (Accession # D4A5M2-1 (CD3E) & P19377-1 (CD3D)).;Rat CD3E&CD3D Heterodimer Protein, His Tag&Flag Tag is produced by co-expression of CD3E and CD3D, has a calculated MW of 14.9 kDa (CD3E) and 14.5 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3D is fused with a flag tag at the C-terminus. As a result of glycosylation, the heterodimer protein migrates as 19-20 kDa under reducing (R) condition, and 40-45 kDa under non-reducing (NR) condition (SDS-PAGE).