Learn More
BIOHIPPO Rat CD30L Microsample
SDP
Catalog No. 5000313746
Encompass
Rat CD30L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD30L in serum plasma tissue homogenates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 49 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD30L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD30L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD30L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13746 Supplier BIOHIPPO Supplier No. BHE15210786
Rat CD30L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD30L in serum plasma tissue homogenates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 49 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD30L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD30L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD30L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color