Learn More
Sino Biological Rat CD36 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80263R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD36 (NP_113749.2)(Gly30-Lys439) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1973