Learn More
Sino Biological Recombinant Rat CD3 epsilon/CD3e Protein (hFc Tag)
Supplier:  Sino Biological 80452R02H100
Encompass_Preferred
A DNA sequence encoding the rat Cd3e (NP_001101610.1) (Tyr24-Asp103) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-228-2301