Learn More
BIOHIPPO Rat CD4 Microsample
SDP
Catalog No. 5000315809
Encompass
Rat CD4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD4 in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 38 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15809 Supplier BIOHIPPO Supplier No. BHE15210525
Rat CD4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD4 in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 38 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CD4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CD4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CD4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color