Learn More
Sino Biological Anti-CD40/TNFRSF5 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 80151MM01A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD40/TNFRSF5 (rR CD40/TNFRSF5; Catalog#80151-R08H; NP_599187.1; Met1-Arg193) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-1462