Learn More
Sino Biological Anti-CD40/TNFRSF5 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80151MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD40 / TNFRSF5 (rR CD40 / TNFRSF5; Catalog#80151-R08H; NP_599187.1; Met1-Arg193). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8205