Learn More
Sino Biological Rat CD48 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80286R08H100
Encompass_Preferred
A DNA sequence encoding the rat CD48 (P10252) (Met1-Arg216) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3732