Learn More
Sino Biological Recombinant Anti-CD5 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80374R006100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD5 / Cluster of Differentiation 5 (rR CD5 / Cluster of Differentiation 5; Catalog#80374-R08H; P51882; Met1-Pro368).
Catalog No. 50-241-1868