Learn More
Sino Biological Anti-CD5 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 80374MM01F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD5 / Cluster of Differentiation 5 (rR CD5 / Cluster of Differentiation 5; Catalog#80374-R08H; P51882; Met1-Pro368) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8851