Learn More
Sino Biological Anti-CD5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80374RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD5 / Cluster of Differentiation 5 (Catalog#80374-R08H; P51882; Met1-Pro368). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8341